Schwefelsäure 96%, reinst
Kst.-Flasche, 1 L
- :7664-93-9
- :H₂SO₄
- :MFCD00064589
- :98.08 g/mol
- :14,08974
- :∼335 °C (1013 hPa)
- :10 °C
- :1,83 g/cm³
- :Raumtemperatur
- :231-639-5
- :8,II
- :Density: 1.84 g/cm³ (20 °C)
- :1830
Documents
- Appearance:Clear liquid
- Color scale:≤10 APHA
- Titration with NaOH:95 to 98 %
- Chloride (Cl):≤0.5 ppm
- Arsenic (As):≤0.03 ppm
- Iron (Fe):≤0.5 ppm
- Heavy metals (as Pb):≤2 ppm
- Packung:Kst.-Flasche
- VE:1 L
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



