Phenoxyethanol 99%
Metalltrommel, 25 L
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: ω-Hydroxyphenetole, β-Hydroxyphenetole, Phenyl cellosolve, Phenoxyethanol, Ethylene glycol monophenyl ether
- CAS Number:122-99-6
- Formula:C₆H₅OCH₂CH₂OH
- MDL Number:MFCD00002857
- Molecular Weight:138.17 g/mol
- Merck Index:12,07410
- Boiling Pt:237 °C (760 mmHg)
- Melting Pt:11…13 °C
- Density:1,1 g/cm³
- Flash Pt:130 °C
- Storage Temperature:Raumtemperatur
- EINECS:204-589-7
Documents
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5360 to 1.5400 (20°C, 589 nm)
- GC:≥98.5 %
- pH:9.5 to 11.0 (5% at 25°C)
- Titration with HClO4:≥99 %
- Iron (Fe):≤0.001 %
Specifications
- Packung:Metalltrommel
- VE:25 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



