Phenoxyethanol 99%
Kst.-Trommel, 5 L
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Synonyme: ω-Hydroxyphenetole, β-Hydroxyphenetole, Phenyl cellosolve, Phenoxyethanol, Ethylene glycol monophenyl ether
- CAS Nummer:122-99-6
- Formel:C₆H₅OCH₂CH₂OH
- MDL-Nummer:MFCD00002857
- Molekulargewicht:138.17 g/mol
- Merck-Index:12,07410
- Siedepunkt:237 °C (760 mmHg)
- Schmelzender Platingehalt:11…13 °C
- Dichte:1,1 g/cm³
- Blitz Pt:130 °C
- Lagertemperatur:Raumtemperatur
- EINECS:204-589-7
Dokumente
Technische Daten
Ergebnisse des Spezifikationstests
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5360 to 1.5400 (20°C, 589 nm)
- GC:≥98.5 %
- pH:9.5 to 11.0 (5% at 25°C)
- Titration with HClO4:≥99 %
- Iron (Fe):≤0.001 %
Technische Daten
- Packung:Kst.-Trommel
- VE:5 L
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



