Natriumpyruvat 99+%
Kst.-Flasche, 500 g
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Synonyme: 2-Ketopropionic acid sodium salt, 2-Oxopropionic acid sodium salt, Sodium 2-oxopropanoate, Pyruvic acid sodium salt, α-Ketopropionic acid sodium salt
- CAS Nummer:113-24-6
- Formel:CH₃COCOONa
- MDL-Nummer:MFCD00002586
- Molekulargewicht:110.04 g/mol
- Schmelzender Platingehalt:>300 °C
- Dichte:1,718 g/cm³ (20 °C)
- Lagertemperatur:Kühlschrank
- EINECS:204-024-4
Documents
ACRO13215
SDS
Technische Daten
Ergebnisse des Spezifikationstests
- Appearance:White to light yellow Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤0.5 %
- Heavy metals:≤10 ppm
- Solubility:(10 % in water) Clear colorless to light yellow
- Titration with HClO4:≥99.0 %
- Chloride (Cl):≤20 ppm
- Sulfate (SO4):≤20 ppm
- Arsenic (As):≤1 ppm
Technische Daten
- Packung:Kst.-Flasche
- VE:500 g



