2,3-Pyridindicarbonsäure 99%
Kst.-Flasche, 500 g
Anbieter: Thermo Fisher Scientific
2,3-Pyridindicarbonsäure 99%
Katalognr. ACRO131855000
Anbieter: Thermo Fisher Scientific
CAS Nummer: 89-00-9
Direkt zu
Product Details & Documents
Synonyme: Quinolinic acid
- CAS Nummer:89-00-9
- Formel:C₇H₅NO₄
- MDL-Nummer:MFCD00006295
- Molekulargewicht:167.12 g/mol
- Merck-Index:12,08258
- Siedepunkt:357 °C (1013 hPa)
- Schmelzender Platingehalt:188…190 °C (Zers.)
- Schmelzender Platingehalt:188…190 °C (Zersetzung)
- Lagertemperatur:Raumtemperatur
- EINECS:201-874-8
Dokumente
Technische Daten
Ergebnisse des Spezifikationstests
- Appearance:White to yellow to light beige Crystalline powder
- Infrared spectrum:Conforms
- Titration with NaOH:≥98.5 %
- Phosphate (PO4):≤1.0 ppm
- Sulfate (SO4):≤1.0 ppm
- Silicate (as SiO2):≤0.01 ppm
- Heavy metals (as Pb):≤0.01 ppm
- KMnO4-reducing substance:Passes test
- Conductivity:2.0 x 10-6 ohm-1 cm-1
Technische Daten
- Packung:Kst.-Flasche
- VE:500 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



