Caprylsäure 99%
Metalltrommel, 25 L
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Synonyme: Octanoic acid, n-Octanoic Acid, Octoic acid
- CAS Nummer:124-07-2
- Formel:C₈H₁₆O₂
- MDL-Nummer:MFCD00004429
- Molekulargewicht:144.21 g/mol
- Merck-Index:13,01771
- Siedepunkt:238 °C (760 mmHg)
- Schmelzender Platingehalt:16…16,5 °C
- Dichte:0,91 g/cm³
- Blitz Pt:110 °C
- Lagertemperatur:Raumtemperatur
- EINECS:204-677-5
- UN:3265
- ADR:8,III
Documents
Technische Daten
Ergebnisse des Spezifikationstests
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4260 to 1.4300 (20°C, 589 nm)
- GC:≥98.5 %
Technische Daten
- Packung:Metalltrommel
- VE:25 L
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



