Dithiooxamid 98%
Kst.-Flasche, 25 g
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Synonyme: Ethanebis(thioamide), Rubeanic acid
- CAS Nummer:79-40-3
- Formel:C₂H₄N₂S₂
- MDL-Nummer:MFCD00004941
- Molekulargewicht:120.2 g/mol
- Schmelzender Platingehalt:200 °C (Zers.)
- Schmelzender Platingehalt:200 °C (Zersetzung)
- Lagertemperatur:Raumtemperatur
Documents
Technische Daten
Ergebnisse des Spezifikationstests
- Appearance:Orange to red to brown Powder
- Infrared spectrum:Conforms
- Sulfated ash:≤2 %
- Elemental Analysis:22.7 to 23.9 % (N)
- Barium (Ba):≤0.005 %
- Calcium (Ca):≤0.05 %
- Iron (Fe):≤0.01 %
- Manganese (Mn):≤5 ppm
- Potassium (K):≤0.005 %
- Sodium (Na):≤0.5 %
- Strontium (Sr):≤0.005 %
- Heavy metals (as Pb):≤0.003 %
- Insoluble matter:≤0.02 % (in dilute HCl)
- Loss on ignition:≤2.0 %
- Water-soluble substances:≤0.4 %
Technische Daten
- Packung:Kst.-Flasche
- VE:25 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



