Dithiooxamid 98%
Glasflasche, 5 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Ethanebis(thioamide), Rubeanic acid
- CAS Number:79-40-3
- Formula:C₂H₄N₂S₂
- MDL Number:MFCD00004941
- Molecular Weight:120.2 g/mol
- Melting Pt:200 °C (Zers.)
- Melting Pt:200 °C (Zersetzung)
- Storage Temperature:Raumtemperatur
Specifications
Specification Test Results
- Appearance:Orange to red to brown Powder
- Infrared spectrum:Conforms
- Sulfated ash:≤2 %
- Elemental Analysis:22.7 to 23.9 % (N)
- Barium (Ba):≤0.005 %
- Calcium (Ca):≤0.05 %
- Iron (Fe):≤0.01 %
- Manganese (Mn):≤5 ppm
- Potassium (K):≤0.005 %
- Sodium (Na):≤0.5 %
- Strontium (Sr):≤0.005 %
- Heavy metals (as Pb):≤0.003 %
- Insoluble matter:≤0.02 % (in dilute HCl)
- Loss on ignition:≤2.0 %
- Water-soluble substances:≤0.4 %
Specifications
- Packung:Glasflasche
- VE:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



