Tropin 98%
Glasflasche, 100 g
: endo-8-Methyl-8-azabicyclo[3.2.1]octan-3-ol
- :120-29-6
- :C₈H₁₅NO
- :MFCD00180667
- :141.21 g/mol
- :233 °C (760 mmHg)
- :62…64 °C
- :1,04 g/cm³
- :Kühlschrank
- :204-384-2
Documents
ACRO14056
SDS
- Appearance:White to light yellow Crystalline powder
- Infrared spectrum:Conforms
- Water:≤2 % (K.F.)
- Titration with HClO4:≥97.5 % (On dried substance)
- Packung:Glasflasche
- VE:100 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



