Imidazol 99%
Kst.-Trommel, 10 kg
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Glyoxaline, 1H-Imidazole, Glyoxalamine
- CAS Number:288-32-4
- Formula:C₃H₄N₂
- MDL Number:MFCD00005183
- Molecular Weight:68.08 g/mol
- Merck Index:13,04935
- Boiling Pt:255…256 °C (760 mmHg)
- Melting Pt:86…90 °C
- Density:1,18 g/cm³ (20 °C)
- Flash Pt:145 °C
- Storage Temperature:Raumtemperatur
- EINECS:206-019-2
- UN:3263
- ADR:8,III
Documents
ACRO12202
SDS
Specifications
Specification Test Results
- Appearance:White to light yellow Flakes
- Infrared spectrum:Conforms
- Water:≤0.2 % (K.F.)
- GC:≥98.5 %
Specifications
- Packung:Kst.-Trommel
- VE:10 kg



