Sulfolan 99+%
Glasflasche, 2,5 L
: Tetrahydrothiophene-1,1-dioxide, Tetramethylene sulphone, Tetrahydrothiophene 1,1-dioxide
- :126-33-0
- :C₄H₈O₂S
- :MFCD00005484
- :120.17 g/mol
- :285 °C (760 mmHg)
- :27,4…27.8 °C
- :1,26 g/cm³
- :165 °C
- :Raumtemperatur
- :204-783-1
- :3335
- :9,III
- Appearance:Clear yellow Liquid after melting
- Infrared spectrum:Conforms
- Refractive index:1.4820 to 1.4850 (20°C, 589 nm)
- Water:≤0.3 %
- Sulfated ash:≤0.1 %
- GC:≥99.0 %
- Packung:Glasflasche
- VE:2,5 L
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



