Phthalaldehyd 98+%
Glasflasche, 100 g
: 1,2-Phthalic dicarboxaldehyde, Phthaldialdehyde, o-Phthalaldehyde, OPA, Phthaldicarboxaldehyde, Benzene-1,2-dicarboxaldehyde
- :643-79-8
- :C₈H₆O₂
- :MFCD00003335
- :134.13 g/mol
- :83…84 °C (1hPa)
- :54…57 °C
- :1,13 g/cm³ (20 °C)
- :132 °C
- :Kühlschrank
- :211-402-2
- :2923
- :8,II
- Appearance:Yellow Powder or crystals and/or lumps
- Infrared spectrum:Conforms
- Melting point:54 to 58 °C
- GC:≥98.0 %
- Packung:Glasflasche
- VE:100 g