4-Brombenzonitril 98%
Kst.-Flasche, 50 g
: p-Bromobenzonitrile
- :623-00-7
- :C₇H₄BrN
- :MFCD00001811
- :182.02 g/mol
- :235…237 °C (760 mmHg)
- :111…114 °C
- :1,562 g/cm³ (20 °C)
- :130 °C
- :Raumtemperatur
- :210-764-9
- :6.1,III
- Appearance:White to light yellow Powder or flakes
- Infrared spectrum:Conforms
- Melting point:111 to 114 °C
- GC:≥97.5 %
- Packung:Kst.-Flasche
- VE:50 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



