Xylit 99+%
Kst.-Flasche, 500 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: (2R,3r,4S)-Pentane-1,2,3,4,5-pentaol
- CAS Number:87-99-0
- Formula:C₅H₁₂O₅
- MDL Number:MFCD00064292
- Molecular Weight:152.15 g/mol
- Merck Index:13,10140
- Boiling Pt:215…217 °C
- Melting Pt:92…96 °C
- Density:1,52 g/cm³ (20 °C)
- Storage Temperature:Raumtemperatur
- EINECS:201-788-0
Documents
Specifications
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:92 to 96 °C
- Water:≤0.5 %
- Sulfated ash:≤0.5 %
- GC:≤2.0 % Other polyols
- Arsenic (As):≤3 ppm
- Heavy metals (as Pb):≤10 ppm
- Reducing sugars:≤0.2 %
Specifications
- Packung:Kst.-Flasche
- VE:500 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



