FastPrep® Empty Tubes and Caps
Screw cap with O-ring, orange
Katalognr. ICNA115063002
Anbieter: MP Biomedicals
Direkt zu
Product Details & Documents
These natural, non-skirted empty Fasrprep® tubes made of PP are used in Fastprep system, to hold sample for homogenisation.
Technische Daten
- Bezeichnung:Screw cap with O-ring, orange
- Seq:01