AuftragsverfolgungKontakt
Trypsin | Avantor
Trypsin
Trypsin-EDTA (recombinant) 1X in PBS, without calcium, without magnesium, without phenol red, 100 ml
Katalognr. L0941-100
Anbieter:   Biowest
Trypsin
Katalognr. L0941-100
Anbieter:  Biowest
CAS Nummer:  
Trypsin
Trypsin
Trypsin-EDTA (recombinant) 1X in PBS, without calcium, without magnesium, without phenol red, 100 ml
CAS Nummer:  
Preis
SFr. 48.60
Einloggen für Verfügbarkeit
1 * 100 mL
Product Details & Documents
Trypsin is a porcine pancreas-derived enzyme that is commonly used for the dissociation and disaggregation of anchorage-dependent mammalian cells and tissues. The concentration of trypsin necessary to dislodge the cells from their substrate depends on the sensitivity of the cells.
Technische Daten
Technische Daten
  • VE:
    100 ml
  • Bezeichnung:
    Trypsin-EDTA (recombinant) 1X in PBS, without calcium, without magnesium, without phenol red
Technische Daten