Phase separating filters
108 H, 125 mm
Catalog # PRATA125108H
Supplier: PRAT DUMAS
Jump to:
Product Details & Documents
Hydrophobic paper for the fast separation of aqueous solutions from hydrofuge solvents.
Useful as well with organic solvents heavier or lighter than water replacing many times bulbs to be settled.
Delivery information: Filters supplied as flat in boxes with 100 filters or folded in 4 by 100.
Specifications
- Sorte:108 H
- Seq:03
- Größe:125 mm