AuftragsverfolgungKontakt
Drum spill kits | Avantor
Drum spill kits
Lab pack, maintenance, SKA-20
Katalognr. BRDY813870
Anbieter:   Brady
Drum spill kits
Katalognr. BRDY813870
Anbieter:  Brady
Drum spill kits
Drum spill kits
Lab pack, maintenance, SKA-20
Preis
SFr. 512.00
Einloggen für Verfügbarkeit
1 * 1 KIT
Product Details & Documents
The drum spill kit is designed for medium sized spills.
  • Water and chemical resistant drum protects sorbents
  • Protects sorbents against moisture, dirt and damage
  • Drum can also be used as shipping container for used sorbents
  • Easy access to products with screw top lid
Lieferinformationen: Supplied with 12 pads (410×510 mm), 3 socs (Ø 7,6×3660 mm), 2 pillows (430×480 mm), 1 pair of gloves, 1 pair of goggles and 3 disposal bags.
Technische Daten
  • Bezeichnung:
    Lab pack, maintenance, SKA-20
  • Seq:
    02