Drum spill kits
Lab pack, oil only, SKO-20
Katalognr. BRDY813869
Anbieter: Brady
Direkt zu
Product Details & Documents
The drum spill kit is designed for medium sized spills.
- Water and chemical resistant drum protects sorbents
- Protects sorbents against moisture, dirt and damage
- Drum can also be used as shipping container for used sorbents
- Easy access to products with screw top lid
Lieferinformationen: Supplied with 12 pads (410×510 mm), 3 socs (Ø 7,6×3660 mm), 2 pillows (430×480 mm), 1 pair of gloves, 1 pair of goggles and 3 disposal bags.
Technische Daten
- Bezeichnung:Lab pack, oil only, SKO-20
- Seq:03