Hygromycin B
Bottle
Katalognr. GENORC-180
Anbieter: G-Biosciences
Direkt zu
Product Details & Documents
Mode of action and Anti-microbial spectrum: Hygromycin B is an aminocyclitol with a wide spectrum of activity that kills bacteria, fungi and higher eukaryotic cells by inhibiting protein synthesis. It is produced by the bacterium Streptomyces hygroscopicus.
- Molecular Biology Grade
- Tested against both sensitive and resistant cells
Synonyme: (3aS,3'R,4S,4'S,5'R,6R,6'R,7S,7aS)-4-(((1R,2S,3R,5S,6R)-3-Amino-2,6-dihydroxy-5-(methylamino)cyclohexyl)oxy)-6'-((S)-1-amino-2-hydroxyethyl)-6-(hydroxymethyl)octahydro-3aH-spiro[[1,3]dioxolo[4,5-c]pyran-2,2'-pyran]-3',4',5',7-tetraol
- CAS Nummer:31282-04-9
- Formel:C₂₀H₃₇N₃O₁₃
- Molekulargewicht:527,6 g/mol
Technische Daten
Technische Daten
- Packung:Bottle
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



