Potassium chloride standard solution
Potassium chloride standard solution, 745,5 mg/ml (0,01 M) KCl, 1412 µS/cm, 50 ml
Katalognr. HACH2974326
Anbieter: Hach
Direkt zu
Product Details & Documents
For calibrations/accuracy checks on conductivity/salinity meters.
Dokumente
Zertifikate
Suche nach ChargennummerTechnische Daten
Technische Daten
- VE:50 ml
- Bezeichnung:Potassium chloride standard solution, 745,5 mg/ml (0,01 M) KCl
- Leitfähigkeit:1412 µS/cm
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.