Dichtungen
Wash seal gasket, 1050, 1100, 1120, 1200, 1220, 1260, 5062-2484, AGLS
Katalognr. CTSA11-3321
Anbieter: SCIENCIX
Direkt zu
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Sciencix parts are equivalent to the corresponding OEM part.
Technische Daten
- Bezeichnung:Wash seal gasket
- Seq:67
- Erstausrüster:AGLS
- Erstausrüster-Äquivalent:5062-2484
- Modell:1050, 1100, 1120, 1200, 1220, 1260