Ersatzteile-Sets
Acquity BSM performance maintenance kit, ACQUITY UPLC BSM, 201000173, WATE
Katalognr. CTSACTS-11091
Anbieter: SCIENCIX
Direkt zu
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Sciencix parts are equivalent to the corresponding OEM part.
Technische Daten
- Bezeichnung:Acquity BSM performance maintenance kit
- Seq:13
- Erstausrüster:WATE
- Erstausrüster-Äquivalent:201000173
- Modell:ACQUITY UPLC BSM