AuftragsverfolgungKontakt
Tris base buffer, powder, Corning® | Avantor
Tris base buffer, powder, Corning®
Tris base buffer, powder, 5.000 g
Katalognr. MDTC61-233-RT
Anbieter:   Corning
Tris base buffer, powder, Corning®
Katalognr. MDTC61-233-RT
Anbieter:  Corning
CAS Nummer:  
Tris base buffer, powder, Corning®
Tris base buffer, powder, Corning®
Tris base buffer, powder, 5.000 g
CAS Nummer:  
Preis
SFr. 896.00
Einloggen für Verfügbarkeit
1 * 5.000 g
Product Details & Documents
Tris is commonly used in buffer solutions in biochemical and molecular biological research.
This primary amine has an effective pH range of 7,0 to 9,2 and is reported to inhibit a number of different enzymes, limiting its usefulness in protein studies to some extent. This tris base buffer is provided in powder form.
Technische Daten
Technische Daten
  • VE:
    5.000 g
  • Bezeichnung:
    Tris base buffer, powder
  • Cat. No.:
    MDTC61-233-RT
  • Lagertemperatur:
    Ambient
  • Transporttemperatur:
    Ambient
Technische Daten