Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Accessories for transilluminators
Accessories for transilluminators
UV-Röhrchen, 8 W, 254 nm, Länge 302 mm
Catalog # CONSVLT8-C
Supplier:   CONSORT
Accessories for transilluminators
Catalog # CONSVLT8-C
Supplier:  CONSORT
Accessories for transilluminators
Accessories for transilluminators
UV-Röhrchen, 8 W, 254 nm, Länge 302 mm
Price
SFr. 109.00
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Bezeichnung:
    UV-Röhrchen, 8 W, 254 nm, Länge 302 mm
  • Seq:
    05
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.