Column installation pre-swaging tools
GC column installation pre-swaging tool, for metal ferrules
HEWLG3440-80218
Handy stainless steel tools for proper column installation lengths into the most popular GC inlets.
- Available for Agilent 7890/6890 model Split/Splitless, MultiMode and Purged Packed inlets
- Each tool has a fitting specific for its ferrules type to provide optimal sealing; one for graphite ferrules, one for metal ferrules
- Easy to read markings for Split/Splitless, MultiMode and Purged Packed inlet lengths
- A ferrule release hole on the Pre-swaging tool for Metal ferrules is insurance against over tightening resulting in a stuck ferrule
Optimize GC performance and reproducibility by ensuring a proper length of column into the inlet. For accurate results, the proper length of column must be inserted into the inlet for optimal sample transfer from the inlet to the column. For consistent results, this process must be done the same every time. Deviating from that length alters results.
- Bezeichnung:GC column installation pre-swaging tool, for metal ferrules
- Seq:02