Preprinted labels, UN3373
Pre-printed labels, Weiß, 110×99 mm
DAKL461846
These preprinted labels are 100% compliant with the UN3373 packaging laws.
- Avoid costly mistakes
- Applied to all diagnostic samples being sent with couriers or regular mail
- Farbe:Weiß
- Bezeichnung:Pre-printed labels
- Abmessungen:110×99 mm
- Seq:02
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



