Ampicillin Trihydrat
500 g
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Ampicillin is lactam antibiotic with an amino group side chain attached to the penicillin structure.
Commonly used to select for ampicillin resistance in mutated and transformed cells
White powder
Soluble in water. Practically insoluble in ethanol
Synonyme: (2S,5R,6R)-6-((R)-2-Amino-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid trihydrate
- CAS Nummer:7177-48-2
- Formel:C₁₆H₁₉N₃O₄S·3H₂O
- MDL-Nummer:MFCD00072036
- Molekulargewicht:403.45 g/mol
- Siedepunkt:684 °C
- Schmelzender Platingehalt:208 °C
- Blitz Pt:>110 °C (230 °F)
- Lagertemperatur:Kühlschrank
- EINECS:200-709-7
Dokumente
Technische Daten
Technische Daten
- VE:500 g



