Sulphadoxin 98%
5 g
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Purity: 98% CAS No.: 2447-57-6 Molecular Formula: C12H14N4O4S Formula Weight: 310.33 Physical Form: Powder Appearance: White Solubility: Soluble in ethanol & ammonium hydroxide (9:1) at 20mg/ml Beilstein Reference: 625453 MDL No.: MFCD00792890
Synonyme: sulphadoxine, 4-Amino-N-(5,6-dimethoxy-4-pyrimidinyl)benzenesulphonamide, 4-Amino-N-(5,6-dimethoxypyrimidin-4-yl)benzenesulphonamide
- CAS Nummer:2447-57-6
- Formel:C₁₂H₁₄N₄O₄S
- MDL-Nummer:MFCD00792890
- Molekulargewicht:310.33 g/mol
- Merck-Index:14,8906
- Lagertemperatur:Raumtemperatur
- EINECS:219-504-9
Dokumente
Technische Daten
Technische Daten
- VE:5 g



