Ampicillin Trihydrat
25 g
Ampicillin is lactam antibiotic with an amino group side chain attached to the penicillin structure.
Commonly used to select for ampicillin resistance in mutated and transformed cells
White powder
Soluble in water. Practically insoluble in ethanol
: (2S,5R,6R)-6-((R)-2-Amino-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid trihydrate
- :7177-48-2
- :C₁₆H₁₉N₃O₄S·3H₂O
- :MFCD00072036
- :403.45 g/mol
- :684 °C
- :208 °C
- :>110 °C (230 °F)
- :Kühlschrank
- :200-709-7
- VE:25 g