Chicken serum
Hühnerserum, hitzeinaktiviert, Sterilfiltriert, gefroren
Katalognr. S050H-1000
Anbieter: Biowest
Direkt zu
Product Details & Documents
Chicken serum is the liquid fraction of whole blood that is collected after the blood is allowed to clot; it is the blood plasma with the fibrinogens removed. Serum includes all proteins not used in blood clotting (coagulation) and all the electrolytes, antibodies, antigens, hormones, and any exogenous substances. Chicken serum is used as a blocking agent in immunoassays and controls.
Dokumente
Zertifikate
Suche nach ChargennummerTechnische Daten
- Serumformat:gefroren
- Sterilität:Sterilfiltriert
- Bezeichnung:Hühnerserum, hitzeinaktiviert
- Seq:18