Rat serum
Rattenserum, Sterilfiltriert, gefroren
Catalog # S2150-500
Supplier: Biowest
Jump to:
Product Details & Documents
Rat serum is the liquid fraction of whole blood that is collected after the blood is allowed to clot; it is the blood plasma with the fibrinogens removed. Serum includes all proteins not used in blood clotting (coagulation) and all the electrolytes, antibodies, antigens, hormones, and any exogenous substances. Rat serum is used as a blocking agent in immunoassays and controls.
Specifications
- Serumformat:gefroren
- Sterilität:Sterilfiltriert
- Bezeichnung:Rattenserum
- Seq:57
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.