Natriumhydroxid NF, Multikompendial, J.T.Baker®
Polyethylene pail, screw top, 12 kg
Supplier: Avantor
Natriumhydroxid NF, Multikompendial, J.T.Baker®
Catalog # 3718-37
Supplier: Avantor
CAS Number: 1310-73-2
Jump to:
Product Details & Documents
Synonyms: Caustic soda
- CAS Number:1310-73-2
- Formula:NaOH
- MDL Number:MFCD00003548
- Molecular Weight:40 g/mol
- Merck Index:14,08627
- Boiling Pt:1390 °C (1013 hPa)
- Melting Pt:323 °C
- Density:2,13 g/cm³ (20 °C)
- Storage Temperature:Raumtemperatur
- EINECS:215-185-5
- UN:1823
- ADR:8,II
- REACH:01-2119457892-27
Specifications
Specification Test Results
- Appearance of solution:passes test
- Assay:97.0 - 100.5 %
- Carbonates:max. 2.0 %
- Chlorides:max. 200 ppm
- Heavy Metals:max. 20 ppm
- Identification A:passes test
- Identification B:passes test
- Iron:max. 10 ppm
- Sulphates:max. 200 ppm
- Chloride:max. 0.050 %
- Clarity and Color of Solution:passes test
- Identification 1:passes test
- Identification 2:passes test
- Mercury:passes test
- Potassium:passes test
- Sodiumcarbonate:max. 2.0 %
- Identification:passes test
- Insoluble substances and organic matters:passes test
Specifications
- VE:12 kg
- Pk Info:Polyethylene pail, screw top
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



