Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Human MCP1 (from E. coli)

Human MCP1 (from E. coli)

20 µG
Catalog # BIRBORB80140-20
Marketsource
Supplier:   Biorbyt
Human MCP1 (from E. coli)
Catalog # BIRBORB80140-20
Supplier:  Biorbyt
Marketsource
CAS Number:  
Human MCP1 (from E. coli)

20 µG

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 20 µG
Product Details & Documents

Documents

Specifications

Specifications

  • VE:
    20 µG
  • Zusammensetzung:
    The protein was lyophilized from concentrated (1mg/ml) sterile solution containing no additives.
  • Cat. No.:
    BIRBORB80140-20
  • Ursprung:
    E. coli
  • Validierte Anwendungen:
    SDS-PAGE , HPLC , FuncS
  • Purity:
    > 98.0% as determined by RP-HPLC and analysis by SDS-PAGE
  • Spezies:
    Human
  • Lagerbedingungen:
    Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
  • Sequence:
    The sequence of the first five N-terminal amino acids was determined and was found to be Gln-Pro-Asp-Ala-Ile.
  • Protein/Peptid-Name:
    MCP1
  • Biologische Aktivität:
    The specific activity as determined by the ability of MCP-1 to chemoattract human Monocytes using concentration of 5-20 ng/ml.
  • Proteinsynonym:
    C-C motif chemokine 2
  • Löslichkeit:
    It is recommended to reconstitute the lyophilized Monocyte Chemotactic Protein-1in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.