Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Split/splitless inlet seals
Split/splitless inlet seals
Seal, gold plated, ultra inert, with washer
Katalognr. HEWL5190-6144
Split/splitless inlet seals
Katalognr. HEWL5190-6144
Split/splitless inlet seals
Split/splitless inlet seals
Seal, gold plated, ultra inert, with washer
Preis
Preis anfragen
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
These inlet seals have been developed to ensure that you have a consistent and inert surface to properly seal the inlet and prevent sample degradation.
  • Very reproducible smooth surface, eliminating traces of machining grooves that can be the source of minute leaks
  • High quality, leak-free seal
Informazioni per l'ordine: An inlet washer is provided with each inlet seal.
Technische Daten
  • Bezeichnung:
    Seal, gold plated, ultra inert, with washer
  • Seq:
    04
Consigli
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..