Ballonflaschen, Biotainer®, Nalgene®
5 l
Katalognr. NALG3405-99
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
PC, clear, with white screw cap.
- Silicone-lined polypropylene screw cap
- Good chemical resistance
- Good temperature resistance
Reduce the risk of carry-over contamination and eliminate the cost of cleaning, sterilizing and associated validations. Use sterile, ready-to-use Biotainer™ Carboys, designed specifically for pharmaceutical and biological manufacturers. Excellent for freezing applications.
Dokumente
Zertifikate
Suche nach ChargennummerCoC_NALG3405-16_1280994 coc_NALG3405-16_1406876 CoC_NALG3405-42_1249486 CoC_NALG3405-42_1258779 CoC_NALG3405-42_1277137
Mehr
NALG3410-42
Bilder 360
NALG3410-42_TDS_EN NALG3423-42_TDS_EN
Technisches Datenblatt
Technische Daten
- Kapazität:5 l