Wafer casettes
4 inch wafer cassette, Edelstahl
Katalognr. PALBPCPC0101
Anbieter: Palbam
Direkt zu
Product Details & Documents
The palbam class stainless steel wafer cassette is designed to replace standard plastic cassettes, in the semiconductor manufacturing process. The cassette has been designed with H Bar.
- Non-contaminating
- Non-deforming
- Fully conductive
- Withstands temperatures up to 400 ºC
- Designed for both manual and robotic handling
Dokumente
Palbam Class - Catalog 2018
Produktliteratur
Technische Daten
- Bezeichnung:4 inch wafer cassette
- Material:Edelstahl
- Seq:01
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.