Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Spray nozzles | Avantor
Spray nozzles
Nozzle for spray gun
Katalognr. ENTE215-15
Anbieter:   ENTEGRIS
Spray nozzles
Katalognr. ENTE215-15
Anbieter:  ENTEGRIS
Spray nozzles
Nozzle for spray gun
Preis
Preis anfragen
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
PFA spray nozzles with NPT connections for use in applications needing chemical or DI water to be sprayed across a surface.
Molded PFA provides universal chemical compatibility and use in ultrapure and chemically corrosive applications.
Technische Daten
  • Bezeichnung:
    Nozzle for spray gun
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.