Wall-mounted cabinets
Wall-mounted cabinet, with one shelf, lockable, 2 sliding glass doors, 1500×350×780 mm
Katalognr. KOTT307-00029
Anbieter: KOETTERMANN
Direkt zu
Product Details & Documents
Made of epoxy powder coated steel, with one shelf.
- Solid hinged doors or sliding glass doors
- Height adjustable shelfs
- Maximum loading capacity: 80 kg
Technische Daten
- Bezeichnung:Wall-mounted cabinet, with one shelf, lockable
- Seq:33
- B×T×H:1500×350×780 mm
- Türen:2 sliding glass doors