Wall-mounted cabinets
Wall- mounted cabinet, with one shelf, open front, -, 600×350×630 mm
Katalognr. KOTT307-00054
Anbieter: KOETTERMANN
Direkt zu
Product Details & Documents
Made of epoxy powder coated steel, with one shelf.
- Solid hinged doors or sliding glass doors
- Height adjustable shelfs
- Maximum loading capacity: 80 kg
Technische Daten
- Bezeichnung:Wall- mounted cabinet, with one shelf, open front
- Seq:20
- B×T×H:600×350×630 mm
- Türen:-