Mobile storage cabinets
Mobile storage cabinet, lockable, 4 drawers, 600×516×740 mm
Katalognr. KOTT315-00076
Anbieter: KOETTERMANN
Direkt zu
Product Details & Documents
Epoxy powder coated steel cabinets
- Optional locks
- Fitted with castors
- Removable shelves
Technische Daten
- Bezeichnung:Mobile storage cabinet, lockable
- Seq:20
- B×T×H:600×516×740 mm
- Türen:4 drawers