Mobile storage cabinets
Mobile storage cabinet with 1 shelf, 1 door, right hinged, 600×516×740 mm
Katalognr. KOTT315-00071
Anbieter: KOETTERMANN
Direkt zu
Product Details & Documents
Epoxy powder coated steel cabinets
- Optional locks
- Fitted with castors
- Removable shelves
Technische Daten
- Bezeichnung:Mobile storage cabinet with 1 shelf
- Seq:15
- B×T×H:600×516×740 mm
- Türen:1 door, right hinged