Mobile storage cabinets
Mobile storage cabinet with 1 shelf, lockable, 1 door, left hinged, 600×516×740 mm
Katalognr. KOTT315-00068
Anbieter: KOETTERMANN
Direkt zu
Product Details & Documents
Epoxy powder coated steel cabinets
- Optional locks
- Fitted with castors
- Removable shelves
Technische Daten
- Bezeichnung:Mobile storage cabinet with 1 shelf, lockable
- Seq:12
- B×T×H:600×516×740 mm
- Türen:1 door, left hinged
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.