Mobile storage cabinets
Mobile storage cabinet, lockable, 1 door, left hinged, 600×516×590 mm
Catalog # KOTT315-00060
Supplier: KOETTERMANN
Jump to:
Product Details & Documents
Epoxy powder coated steel cabinets
- Optional locks
- Fitted with castors
- Removable shelves
Specifications
- Bezeichnung:Mobile storage cabinet, lockable
- Seq:04
- B×T×H:600×516×590 mm
- Türen:1 door, left hinged