Mobile storage cabinets
Mobile storage cabinet, 1 door, left hinged, 600×516×590 mm
Katalognr. KOTT315-00059
Anbieter: KOETTERMANN
Direkt zu
Product Details & Documents
Epoxy powder coated steel cabinets
- Optional locks
- Fitted with castors
- Removable shelves
Technische Daten
- Bezeichnung:Mobile storage cabinet
- Seq:03
- B×T×H:600×516×590 mm
- Türen:1 door, left hinged