Mobile storage cabinets
Mobile storage cabinet, with 2 drawers of 150 mm height and 1 drawer of 300 mm height, lockable, 3 drawers, 600×516×590 mm
Katalognr. KOTT315-00058
Anbieter: KOETTERMANN
Direkt zu
Product Details & Documents
Epoxy powder coated steel cabinets
- Optional locks
- Fitted with castors
- Removable shelves
Technische Daten
- Bezeichnung:Mobile storage cabinet, with 2 drawers of 150 mm height and 1 drawer of 300 mm height, lockable
- Seq:02
- B×T×H:600×516×590 mm
- Türen:3 drawers



