Mobile storage cabinets
Mobile storage cabinet with 1 shelf, With front open, 600×500×590 mm
KOTT315-00057
Epoxy powder coated steel cabinets
- Optional locks
- Fitted with castors
- Removable shelves
- Bezeichnung:Mobile storage cabinet with 1 shelf
- Seq:01
- B×T×H:600×500×590 mm
- Türen:With front open