Bunsen burners, LabFlame
LabFlame safety burner with IR-sensor and LC display, All kind of gas sources
Catalog # BTOLLF8.200.000
Supplier: BioTool
Jump to:
Product Details & Documents
Stainless steel Bunsen burners, with removable burner head. The optimised air-flow system allows a uniform and constant flame even in safety cabinets with horizontal or vertical air flow. All controls for gas and air supply as well as the multifunctional control for the modes of operation are conveniently located on the easy to read display.
- Can be connected to any kind of gas source
- Optimised for use in microbiology laboratories
- Extremely firm and consistent flame produced with only a few flicks of the wrist
Documents
Specifications
- Bezeichnung:LabFlame safety burner with IR-sensor and LC display
- Seq:02
- Für:All kind of gas sources