Echtgrün FCF
Glasflasche für Feststoffe, 5 g
Katalognr. CAYM14335-5
Anbieter: Cayman Chemical
Direkt zu
Product Details & Documents
Synonyme: Light green, Sodium 2-((4-(ethyl(3-sulphonatobenzyl)amino)phenyl)(4-(ethyl(3-sulphonatobenzyl)iminio)cyclohexa-2,5-dien-1-ylidene)methyl)-5-hydroxybenzenesulphonate, CI 42053, Dihydrogen (ethyl)[4-[4-[ethyl(3-sulphonatobenzyl)amino](4-hydroxy-2-sulphonatobenzhydrylidene]cyclohexa-2,5-dien-1-ylidene](3-sulphonatobenzyl)ammonium, disodium salt
- CAS Nummer:2353-45-9
- Formel:C₃₇H₃₄N₂Na₂O₁₀S₃
- MDL-Nummer:MFCD00013053
- Molekulargewicht:808.86 g/mol
- Merck-Index:13,03970
- Schmelzender Platingehalt:290 °C
- Lagertemperatur:Raumtemperatur
- EINECS:219-091-5
Technische Daten
Technische Daten
- Packung:Glasflasche für Feststoffe
- VE:5 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



