Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Virginiamycin complex ≥95% | Avantor

Virginiamycin complex ≥95%

5 mg
Katalognr. CAYM14503-5
Anbieter:   Cayman Chemical
Virginiamycin complex ≥95%
Katalognr. CAYM14503-5
Anbieter:  Cayman Chemical
CAS Nummer:  121006-76-1
Virginiamycin complex ≥95%

5 mg

CAS Nummer:  121006-76-1
Preis
SFr. 583.00
Einloggen für Verfügbarkeit
1 * 5 mg
Product Details & Documents

Virginiamycin complex is a complex containing two streptogramin antibiotics, virginiamycin M1 (75%) and virginiamycin S1 (25%), produced by S. virginiae.

As a complex, the two antibiotics act synergistically to irreversibly inhibit protein synthesis in bacteria. When given to livestock, virginiamycin improves growth. Virginiamycin complex is also used in fuel ethanol fermenters to prevent to control bacteria competing with yeast for nutrients.

  • CAS Nummer:121006-76-1
  • Lagertemperatur:Gefrierschrank
Technische Daten

Technische Daten

  • VE:
    5 mg

Technische Daten